Assay Details
| Gene Symbol: RNASE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040590.1 | NP_001035680.1 | 1 - 2 | 128 | 67 | |
| GenBank mRNA | BC102072.1 | - | 1 - 2 | 143 | 67 | |
| BT021551.1 | - | 1 - 2 | 128 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102072.1, NM_001040590.1 transcripts.
Set Membership: |
|

RNASE4
Genomic Map