Assay Details
| Gene Symbol: POFUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043342.2 | NP_001036807.1 | 3 - 4 | 504 | 93 | |
| NM_001043343.1 | NP_001036808.1 | 3 - 4 | 506 | 93 | ||
| NM_001043344.1 | NP_001036809.1 | 3 - 4 | 506 | 93 | ||
| NM_001043345.2 | NP_001036810.1 | 3 - 4 | 504 | 93 | ||
| NM_182986.2 | NP_892031.1 | 3 - 4 | 506 | 93 | ||
| GenBank mRNA | AJ781503.1 | - | 3 - 4 | 438 | 93 | |
| AY344580.2 | - | 3 - 4 | 506 | 93 | ||
| AY344581.2 | - | 3 - 4 | 506 | 93 | ||
| BC126762.1 | - | 3 - 4 | 479 | 93 | ||
| BT021843.1 | - | 3 - 4 | 486 | 93 | ||
| DQ138291.1 | - | 3 - 4 | 506 | 93 | ||
| DQ138292.1 | - | 3 - 4 | 506 | 93 | ||
| DQ138293.1 | - | 3 - 4 | 506 | 93 | ||
More Information
Set Membership: |
|

POFUT1
Genomic Map