Assay Details
| Gene Symbol: NAGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046349.1 | NP_001039814.1 | 9 - 10 | 1229 | 69 | |
| GenBank mRNA | BC114813.1 | - | 9 - 10 | 1255 | 69 | |
| BT021618.1 | - | 9 - 10 | 1229 | 69 | ||
More Information
Set Membership: |
|

NAGA
Genomic Map