Assay Details
| Gene Symbol: TFDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075241.1 | NP_001068709.1 | 6 - 7 | 667 | 66 | |
| GenBank mRNA | BC109872.1 | - | 7 - 8 | 668 | 66 | |
| BT021647.1 | - | 6 - 7 | 667 | 66 | ||
More Information
Set Membership: |
|

TFDP2
Genomic Map