Assay Details
| Gene Symbol: LRRC39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076425.1 | NP_001069893.1 | 8 - 9 | 1079 | 76 | |
| GenBank mRNA | BC102917.1 | - | 8 - 9 | 1079 | 76 | |
More Information
Set Membership: |
|

LRRC39
Genomic Map