Assay Details
| Gene Symbol: GPR89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076970.1 | NP_001070438.1 | 9 - 10 | 952 | 153 | |
| GenBank mRNA | BT021195.1 | - | 9 - 10 | 952 | 153 | |
| BT021606.1 | - | 9 - 10 | 948 | 153 | ||
More Information
Set Membership: |
|

GPR89A
Genomic Map