Assay Details
| Gene Symbol: B3GNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076979.1 | NP_001070447.1 | 2 - 3 | 1614 | 90 | |
| GenBank mRNA | BT021862.1 | - | 2 - 3 | 1614 | 90 | |
More Information
Set Membership: |
|

B3GNT5
Genomic Map