Assay Details
| Gene Symbol: CENPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077003.1 | NP_001070471.1 | 4 - 5 | 609 | 56 | |
| GenBank mRNA | BC103238.1 | - | 4 - 5 | 609 | 56 | |
More Information
Set Membership: |
|

CENPO
Genomic Map