Assay Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077864.1 | NP_001071332.1 | 3 - 4 | 290 | 76 | |
| GenBank mRNA | BC103116.1 | - | 3 - 4 | 290 | 76 | |
More Information
Set Membership: |
|

STX7
Genomic Map