Assay Details
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079793.2 | NP_001073261.1 | 2 - 3 | 246 | 109 | |
GenBank mRNA | BC103392.1 | - | 2 - 3 | 248 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
