Assay Details
| Gene Symbol: SDR42E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080292.1 | NP_001073761.1 | 1 - 2 | 36 | 59 | |
| GenBank mRNA | BC109694.1 | - | 1 - 2 | 61 | 59 | |
| BT021824.1 | - | 1 - 2 | 36 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109694.1, NM_001080292.1 transcripts.
Set Membership: |
|

SDR42E1
Genomic Map