Assay Details
| Gene Symbol: SDR42E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080292.1 | NP_001073761.1 | 2 - 3 | 125 | 67 | |
| GenBank mRNA | BC109694.1 | - | 2 - 3 | 150 | 67 | |
| BT021824.1 | - | 2 - 3 | 125 | 67 | ||
More Information
Set Membership: |
|

SDR42E1
Genomic Map