Assay Details
| Gene Symbol: EMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080327.1 | NP_001073796.1 | 10 - 11 | 816 | 74 | |
| GenBank mRNA | BC102870.1 | - | 10 - 11 | 827 | 74 | |
| BC142076.1 | - | 10 - 11 | 850 | 74 | ||
| BT021027.1 | - | 10 - 11 | 816 | 74 | ||
More Information
Set Membership: |
|

EMC2
Genomic Map