Assay Details
| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080919.1 | NP_001074388.1 | 2 - 3 | 415 | 85 | |
| GenBank mRNA | BT021839.1 | - | 2 - 3 | 415 | 85 | |
More Information
Set Membership: |
|

SULT1C4
Genomic Map