Assay Details
| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080920.1 | NP_001074389.1 | 7 - 8 | 792 | 57 | |
| GenBank mRNA | BT021809.1 | - | 7 - 8 | 792 | 57 | |
More Information
Set Membership: |
|

TSPAN18
Genomic Map