Assay Details
| Gene Symbol: WBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082441.1 | NP_001075910.1 | 3 - 4 | 352 | 125 | |
| GenBank mRNA | AF322215.1 | - | 3 - 4 | 352 | 125 | |
More Information
Set Membership: |
|

WBP2NL
Genomic Map