Assay Details
| Gene Symbol: IL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173924.2 | NP_776349.1 | 4 - 5 | 398 | 64 | |
| GenBank mRNA | AF348422.1 | - | 4 - 5 | 398 | 64 | |
| BC142466.1 | - | 4 - 5 | 497 | 64 | ||
| EU276072.1 | - | 4 - 5 | 397 | 64 | ||
More Information
Set Membership: |
|

IL7
Genomic Map