Assay Details
| Gene Symbol: TXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173968.3 | NP_776393.1 | 1 - 2 | 106 | 71 | |
| GenBank mRNA | AF104105.1 | - | 1 - 2 | 103 | 71 | |
| BC103415.1 | - | 1 - 2 | 106 | 71 | ||
| BC120457.1 | - | 1 - 2 | 105 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103415.1, BC120457.1, NM_173968.3 transcripts.
Set Membership: |
|

TXN
Genomic Map