Assay Details
| Gene Symbol: TXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173968.3 | NP_776393.1 | 2 - 3 | 199 | 67 | |
| GenBank mRNA | AF104105.1 | - | 2 - 3 | 196 | 67 | |
| BC103415.1 | - | 2 - 3 | 199 | 67 | ||
| BC120457.1 | - | 2 - 3 | 198 | 67 | ||
More Information
Set Membership: |
|

TXN
Genomic Map