Assay Details
| Gene Symbol: TXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173968.3 | NP_776393.1 | 4 - 5 | 331 | 128 | |
| GenBank mRNA | AF104105.1 | - | 4 - 5 | 328 | 128 | |
| BC103415.1 | - | 4 - 5 | 331 | 128 | ||
| BC120457.1 | - | 4 - 5 | 330 | 128 | ||
More Information
Set Membership: |
|

TXN
Genomic Map