Assay Details
| Gene Symbol: CALR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174000.2 | NP_776425.1 | 3 - 4 | 464 | 64 | |
| GenBank mRNA | AB067687.1 | - | 3 - 4 | 464 | 64 | |
| BC140582.1 | - | 3 - 4 | 457 | 64 | ||
| L13462.1 | - | 628 | 64 | |||
More Information
Set Membership: |
|

CALR
Genomic Map