Assay Details
| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174014.2 | NP_776439.1 | 3 - 4 | 464 | 67 | |
| GenBank mRNA | AF272828.1 | - | 3 - 4 | 464 | 67 | |
| BC142335.1 | - | 3 - 4 | 472 | 67 | ||
More Information
Set Membership: |
|

CD69
Genomic Map