Assay Details
| Gene Symbol: MYH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174117.1 | NP_776542.1 | 30 - 31 | 4421 | 92 | |
| GenBank mRNA | AB059399.2 | - | 30 - 31 | 4421 | 92 | |
More Information
Set Membership: |
|

MYH1
Genomic Map