Assay Details
| Gene Symbol: PRND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174158.2 | NP_776583.1 | 1 - 2 | 96 | 69 | |
| GenBank mRNA | AJ278011.1 | - | 1 - 2 | 96 | 69 | |
| BC133367.1 | - | 1 - 2 | 58 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ278011.1, BC133367.1, NM_174158.2 transcripts.
Set Membership: |
|

PRND
Genomic Map