Assay Details
| Gene Symbol: CCL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174263.2 | NP_776688.1 | 1 - 2 | 115 | 79 | |
| GenBank mRNA | AY061959.1 | - | 1 - 2 | 115 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCL20| Gene Symbol: CCL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174263.2 | NP_776688.1 | 1 - 2 | 115 | 79 | |
| GenBank mRNA | AY061959.1 | - | 1 - 2 | 115 | 79 | |
Set Membership: |
|