Assay Details
| Gene Symbol: CRYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174293.2 | NP_776718.1 | 6 - 7 | 794 | 60 | |
| GenBank mRNA | AF480862.1 | - | 6 - 7 | 794 | 60 | |
| BC140654.1 | - | 6 - 7 | 796 | 60 | ||
More Information
Set Membership: |
|

CRYL1
Genomic Map