Assay Details
| Gene Symbol: GNGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174327.3 | NP_776752.1 | 2 - 3 | 165 | 87 | |
| GenBank mRNA | K02199.1 | - | 1 - 2 | 156 | 87 | |
| K03255.1 | - | 2 - 3 | 165 | 87 | ||
More Information
Set Membership: |
|

GNGT1
Genomic Map