Assay Details
| Gene Symbol: HINT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174340.2 | NP_776765.1 | 3 - 4 | 389 | 65 | |
| GenBank mRNA | AF366940.1 | - | 3 - 4 | 389 | 65 | |
| BC114199.1 | - | 3 - 4 | 360 | 65 | ||
More Information
Set Membership: |
|

HINT2
Genomic Map