Assay Details
| Gene Symbol: IL1RN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174357.3 | NP_776782.1 | 2 - 3 | 221 | 88 | |
| GenBank mRNA | AB005148.1 | - | 2 - 3 | 223 | 88 | |
| BC134577.1 | - | 3 - 4 | 202 | 88 | ||
More Information
Set Membership: |
|

IL1RN
Genomic Map