Assay Details
| Gene Symbol: KLRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174376.2 | NP_776801.1 | 1 - 2 | 143 | 93 | |
| GenBank mRNA | AY075101.1 | - | 1 - 2 | 143 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174376.2 transcripts.
Set Membership: |
|

KLRA1
Genomic Map