Assay Details
| Gene Symbol: KLRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174376.2 | NP_776801.1 | 7 - 8 | 994 | 91 | |
| GenBank mRNA | AY075101.1 | - | 7 - 8 | 994 | 91 | |
| GQ339253.1 | - | 7 - 8 | 994 | 91 | ||
| GQ339254.1 | - | 7 - 8 | 994 | 91 | ||
More Information
Set Membership: |
|

KLRA1
Genomic Map