Assay Details
| Gene Symbol: LECT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174380.2 | NP_776805.1 | 3 - 4 | 291 | 118 | |
| GenBank mRNA | AB001350.1 | - | 3 - 4 | 291 | 118 | |
| BC103166.1 | - | 3 - 4 | 466 | 118 | ||
| D89011.1 | - | 3 - 4 | 421 | 118 | ||
More Information
Set Membership: |
|

LECT2
Genomic Map