Assay Details
| Gene Symbol: MYOZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174397.2 | NP_776822.1 | 6 - 7 | 854 | 75 | |
| GenBank mRNA | AF359567.1 | - | 6 - 7 | 854 | 75 | |
| BC122689.1 | - | 5 - 6 | 753 | 75 | ||
More Information
Set Membership: |
|

MYOZ1
Genomic Map