Assay Details
| Gene Symbol: YWHAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174491.3 | NP_776916.1 | 3 - 4 | 460 | 80 | |
| GenBank mRNA | AF043735.1 | - | 3 - 4 | 407 | 80 | |
| BC102928.1 | - | 3 - 4 | 460 | 80 | ||
More Information
Set Membership: |
|

YWHAE
Genomic Map