Assay Details
| Gene Symbol: ADORA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174497.3 | NP_776922.2 | 1 - 2 | 859 | 62 | |
| GenBank mRNA | BC142208.1 | - | 1 - 2 | 822 | 62 | |
| BC142265.1 | - | 2 - 3 | 527 | 62 | ||
| BT021759.1 | - | 1 - 2 | 859 | 62 | ||
| M86261.1 | - | 1 - 2 | 829 | 62 | ||
| X63592.1 | - | 367 | 62 | |||
More Information
Set Membership: |
|

ADORA1
Genomic Map