Assay Details
| Gene Symbol: IGFBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174556.1 | NP_776981.1 | 1 - 2 | 541 | 61 | |
| GenBank mRNA | AF305199.1 | - | 1 - 2 | 522 | 61 | |
| BC149336.1 | - | 1 - 2 | 542 | 61 | ||
| M76478.1 | - | 1 - 2 | 524 | 61 | ||
More Information
Set Membership: |
|

IGFBP3
Genomic Map