Assay Details
| Gene Symbol: POU5F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174580.3 | NP_777005.2 | 2 - 3 | 526 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POU5F1| Gene Symbol: POU5F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174580.3 | NP_777005.2 | 2 - 3 | 526 | 75 | |
Set Membership: |
|