Assay Details
| Gene Symbol: PTGER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174588.2 | NP_777013.1 | 1 - 2 | 825 | 102 | |
| GenBank mRNA | AF539402.1 | - | 1 - 2 | 825 | 102 | |
More Information
Set Membership: |
|

PTGER2
Genomic Map