Assay Details
| Gene Symbol: SERPINE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174669.2 | NP_777094.1 | 5 - 6 | 1060 | 61 | |
| GenBank mRNA | AF210380.1 | - | 1 - 2 | 215 | 61 | |
| AF251153.1 | - | 5 - 6 | 1060 | 61 | ||
| AY581202.1 | - | 5 - 6 | 1043 | 61 | ||
| BC123834.1 | - | 5 - 6 | 1055 | 61 | ||
More Information
Set Membership: |
|

SERPINE2
Genomic Map