Assay Details
| Gene Symbol: ATP5PF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174717.3 | NP_777142.1 | 1 - 2 | 46 | 72 | |
| GenBank mRNA | BC103429.1 | - | 1 - 2 | 49 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103429.1, NM_174717.3 transcripts.
Set Membership: |
|

ATP5PF
Genomic Map