Assay Details
| Gene Symbol: VDAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174729.2 | NP_777154.1 | 1 - 2 | 66 | 78 | |
| GenBank mRNA | AF268466.1 | - | 1 - 2 | 66 | 78 | |
| BC146149.1 | - | 2 - 3 | 160 | 78 | ||
More Information
Set Membership: |
|

VDAC3
Genomic Map