Assay Details
| Gene Symbol: KIR2DL5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174740.2 | NP_777165.1 | 6 - 7 | 812 | 74 | |
| GenBank mRNA | AF490399.1 | - | 6 - 7 | 812 | 74 | |
| AY075102.1 | - | 6 - 7 | 812 | 74 | ||
More Information
Set Membership: |
|

KIR2DL5A
Genomic Map