Assay Details
| Gene Symbol: BCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174741.2 | NP_777166.1 | 6 - 7 | 825 | 67 | |
| GenBank mRNA | AF270512.2 | - | 6 - 7 | 825 | 67 | |
| BC123496.1 | - | 6 - 7 | 828 | 67 | ||
More Information
Set Membership: |
|

BCAM
Genomic Map