Assay Details
| Gene Symbol: PTHLH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174753.1 | NP_777178.1 | 3 - 4 | 662 | 86 | |
| GenBank mRNA | AB097837.1 | - | 3 - 4 | 662 | 86 | |
| BC149411.1 | - | 4 - 5 | 650 | 86 | ||
More Information
Set Membership: |
|

PTHLH
Genomic Map