Assay Details
| Gene Symbol: CA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175717.2 | NP_783648.1 | 4 - 5 | 821 | 89 | |
| GenBank mRNA | AY075023.1 | - | 4 - 5 | 821 | 89 | |
| BC123827.1 | - | 4 - 5 | 647 | 89 | ||
More Information
Set Membership: |
|

CA11
Genomic Map