Assay Details
| Gene Symbol: MUC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176631.1 | NP_788804.1 | 2 - 3 | 798 | 94 | |
| GenBank mRNA | AJ417816.1 | - | 2 - 3 | 798 | 94 | |
| BC151701.1 | - | 3 - 4 | 803 | 94 | ||
More Information
Set Membership: |
|

MUC15
Genomic Map