Assay Details
| Gene Symbol: FSCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176633.3 | NP_788806.1 | 3 - 4 | 1141 | 62 | |
| GenBank mRNA | AB003485.1 | - | 3 - 4 | 1143 | 62 | |
More Information
Set Membership: |
|

FSCN2
Genomic Map