Assay Details
| Gene Symbol: ARHGDIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176650.3 | NP_788823.1 | 5 - 6 | 491 | 63 | |
| GenBank mRNA | BC103465.1 | - | 5 - 6 | 491 | 63 | |
| BT021511.1 | - | 5 - 6 | 472 | 63 | ||
| X52689.1 | - | 461 | 63 | |||
More Information
Set Membership: |
|

ARHGDIA
Genomic Map