Assay Details
| Gene Symbol: FDPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177497.2 | NP_803463.1 | 1 - 2 | 145 | 64 | |
| GenBank mRNA | AF461050.1 | - | 1 - 2 | 145 | 64 | |
| BC111256.1 | - | 2 - 3 | 965 | 64 | ||
| BC120218.1 | - | 2 - 3 | 207 | 64 | ||
| BC149572.1 | - | 2 - 3 | 266 | 64 | ||
More Information
Set Membership: |
|

FDPS
Genomic Map