Assay Details
| Gene Symbol: GGTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177511.2 | NP_803477.1 | 8 - 9 | 881 | 117 | |
| GenBank mRNA | BC151691.1 | - | 7 - 8 | 593 | 117 | |
| J04989.1 | - | 8 - 9 | 881 | 117 | ||
More Information
Set Membership: |
|

GGTA1
Genomic Map